Learn More
IBA LifeSciences pASK-IBA6C
Supplier:  IBA LifeSciences 21325000
Encompass_Preferred
pASK-IBA6C is a classic cloning vector devised for E. coli expression of recombinant proteins with the Strep-tagII fused to the N-terminus. It carries the chloramphenicol resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, a multiple cloning site with recognition sites for several common restriction enzymes (e.g. EcoRI, SmaI, BamHI, SalI, PstI), the inducible tetracycline promoter/operator for the regulated expression of proteins, the ompA signal for periplasmic secretion of the recombinant protein, and the sequence for a protease cleavage site.Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity. During the secretion into the periplasmic space the ompA signal sequence will be cleaved off. The N-terminal Strep-tagII can be removed by the protease factor Xa.
Catalog No. NC3566328