Learn More
Lonza Walkersville PASMC - Hu Pulmonary Artery Sm
Supplier:  Lonza Walkersville CC2581
Encompass
Human Pulmonary Artery Smooth Muscle Cells (PASMC) are shipped in 3rd passage Human Pulmonary Artery Smooth Muscle Cells are guaranteed for 15 population doublings and to stain positive for alpha smooth muscle actin and negative for von Willebrand Factor VIII after differentiation All cells test negative for mycoplasma bacteria yeast and fungi HIV-1 hepatitis B and hepatitis C are not detected for all donors and/or cell lots A Certificate of Analysis (CoA) is provided for each cell lot purchased
Catalog No. 50-004-96399