Learn More
Foxx Life Sciences Abdos Pasteur Pipettes, Low Density Polyethylene (LDPE) 1ml, 500/CS
Supplier:  Foxx Life Sciences P31201
Encompass_Preferred
Pasteur Pipettes made of Low-density polyethylene (LDPE), are unbreakable, non-toxic all-in one pipettes. These pipettes eliminate the hazard of broken glass and exposure to infectious materials. Molded from translucent high-quality Polyethylene, these pipettes are inert to biological fluids and most acids.•Can be sealed and refrigerated•The low-affinity low binding surface reduces the loss of cells and valuable proteins•Sterile and non-sterile options availableFoxx Life Sciences is proud to represent high-quality Abdos plasticware and plastic laboratory products. For downloadable Certificates please visit: http://www.abdoslifesciences.com/Global/certificates.html.
Catalog No. NC1819504