Learn More
Cole-Parmer Essentials Plastic Pasteur Pipettes, 5.8" Length, Sterile; 500/CS
Supplier:  Cole-Parmer 3456790
Encompass_Preferred
Constructed of polystyrene, these transfer pasteur pipettes won't shatter like glass — no need for a Sharps container.
Catalog No. NC2007315