Learn More
Andwin Scientific PASTEUR PIPETTE RUBBERBULB 2ML
SDP
Supplier:  Andwin Scientific Z111597
Encompass_Preferred
The pasteur rubber bulb features thin walls and small size that can be used with small pipets. This product has a volume capacity of 2 mL, a bore size of 6 mm, and a length of 57 mm.
Catalog No. NC9894128