Learn More
Enzo Life Sciences Patulin (25mg). CAS: 149-29-1
SDP
Supplier:  Enzo Life Sciences ALX270111M025
Encompass_Preferred
Inhibitor of protein farnesylation in a cell free assay. Inhibits incorporation of tritiated mevalonate into proteins in whole cells. Mycotoxin with anti-bacterial, potassium uptake inhibitory and possibly carcinogenic activities. Known contaminant of spoiled apples. Alternative name: 4-Hydroxy-4H-furo[3,2-c]pyran-2(6H)-one, Clairformin, Clavacin, Clavatin, Claviformin, Leucopin, Mycoin C3, Mycosin, Penicidin, Terinin. Formula: C7H6O4. MW: 154.1. MI: 14: 7053. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in DMSO, methanol, ethanol or water. Long Term Storage: -20°C. Handling: Protect from light.
Catalog No. 50-200-8411