Learn More
Abcam Patulin, fungal mycoto x in, 1MG
Supplier:  Abcam AB1427251MG
Encompass_Preferred
MW 154.12 Da, Purity >98%. Fungal mycotoxin, DNA damaging agent and induces ROS production. Apoptosis inducer via EGR-1 dependent ATF3 upregulation in vitro. Anti-bacterial, carcinogenic, mutagenic and anticancer agent. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6197