Learn More
Selleck Chemical LLC Pazopanib S3012-1g
SDP
Supplier:  Selleck Chemical LLC S30121G
Encompass_Preferred
Pazopanib (GW786034) is a novel multi-target inhibitor of VEGFR1 VEGFR2 VEGFR3 PDGFR FGFR c-Kit and c-Fms/CSF1R with IC50 of 10 nM 30 nM 47 nM 84 nM 74 nM 140 nM and 146 nM in cell-free assays respectively Pazopanib induces cathepsin B activation and autophagy
Catalog No. 50-310-9382