Learn More
Medchemexpress LLC Pazopanib-d6 | 1219592-01-4 | 98.0% | 443.55 | C21H17D6N7O2S | 500 UG
SDP
Supplier:  Medchemexpress LLC HY10208S500UG
Encompass_Preferred
Pazopanib-d6 is the deuterium-labeled analogue of pazopanib, provided as a high-purity powder for use as a stable isotope internal standard in bioanalytical quantification (e.g., LC-MS). It is intended for research applications requiring accurate measurement of pazopanib concentrations.
  • Deuterium-labeled internal standard for accurate quantification.
  • Suitable for LC-MS and GC-MS bioanalytical assays.
  • High purity (98.0%) to minimize analytical interference.
  • Storage: powder at -20°C for long-term, 4°C for short-term.
  • Available in small research pack sizes, including 500 μg.
Catalog No. 50-002-45518