Learn More
Apexbio Technology LLC Pazopanib (GW-786034), 500mg. Cas: 444731-52-6 MFCD: MFCD11616589. VEGFR/PDGFR/FGFR inhibitor
SDP
Supplier:  Apexbio Technology LLC A3022500
Encompass_Preferred
Pazopanib is a potent and selective second generation multi-targeted receptor tyrosine kinase inhibitor of vascular endothelial growth factor receptor (VEGFR), platelet-derived growth factor receptor (PDGFR) and c-kit tyrosine kinases. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4784