Learn More
Medchemexpress LLC HY-12009 10mg Medchemexpress, Pazopanib (Hydrochloride) CAS:635702-64-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1200910MG
Encompass_Preferred
Medchemexpress, HY-12009 10mg Pazopanib (HCl) CAS:635702-64-6 Pazopanib Hydrochloride (GW786034 Hydrochloride) is a novel multi-target inhibitor of VEGFR1, VEGFR2, VEGFR3, PDGFRβ, c-Kit, FGFR1, and c-Fms with an IC50 of 10, 30, 47, 84, 74, 140 and 146 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9574