Learn More
Medchemexpress LLC Pazufloxacin (mesylate) | 163680-77-1 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0724A10MM/1ML
Encompass_Preferred
Pazufloxacin mesylate is an orally active fluoroquinolone antimicrobial agent. It inhibits DNA gyrase with IC50 values of 0.88 μg/mL (E. coli) and 1.9 μg/mL (P. aeruginosa). This agent exhibits broad-spectrum antimicrobial activity with MIC90 values ranging from 0.025 to 100 μg/mL against Gram-positive and Gram-negative bacteria, non-fermenting bacteria, Legionella spp., and anaerobic bacteria. It is indicated for research on systemic infections, lung infections, urinary tract infections, and Legionella pneumonia.
  • Orally active fluoroquinolone antimicrobial agent
  • Inhibits DNA gyrase
  • Exhibits broad-spectrum antimicrobial activity against a variety of bacteria, including gram-positive, gram-negative, non-fermenting, Legionella spp., and anaerobic bacteria
  • Indicated for research on systemic infections, lung infections, urinary tract infections, and Legionella pneumonia
Catalog No. 50-004-07083