Learn More
Medchemexpress LLC Pazufloxacin (mesylate) | 163680-77-1 | 99.97% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0724A500MG
Encompass_Preferred
Pazufloxacin mesylate is an orally active fluoroquinolone antimicrobial agent with a purity of 99.97%. It effectively inhibits DNA gyrase and exhibits broad-spectrum antimicrobial activity against a wide range of bacteria. This compound is used in research for systemic, lung, and urinary tract infections, as well as Legionella pneumonia.
  • Orally active fluoroquinolone antimicrobial agent
  • Inhibits DNA gyrase
  • Exhibits broad-spectrum antimicrobial activity
  • Suitable for research on systemic, lung, and urinary tract infections
  • Suitable for research on Legionella pneumonia
  • High purity of 99.97%
  • Demonstrates efficacy in vivo against pulmonary infections
  • Reduces bacterial counts in Legionella pneumonia models
Catalog No. 50-004-08187