Learn More
Biolegend Pacific Blue™ anti-human CD19
Supplier:  Biolegend 302232
Encompass_Preferred
Pacific Blue™ anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-165-845