Learn More
Biolegend Pacific Blue™ anti-human Ki-67
Supplier:  Biolegend 350511
Encompass_Preferred
Pacific Blue™ anti-human Ki-67 [Ki-67]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Bovine; Apps: ICFC; Size: 25 tests
Catalog No. 50-168-609