Learn More
Biolegend Pacific Blue™ anti-mouse CD3
Supplier:  Biolegend 100213
Encompass_Preferred
Pacific Blue™ anti-mouse CD3 [17A2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-373