Learn More
Biolegend Pacific Blue™ anti-mouse CD3ε
Supplier:  Biolegend 100333
Encompass_Preferred
Pacific Blue™ anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-162-407