Learn More
Biolegend Pacific Blue™ anti-MU/HU GL7 Antigen (T/B Cell Act. Marker)
Supplier:  Biolegend 144613
Encompass_Preferred
Pacific Blue™ anti-MU/HU GL7 Antigen (T/B Cell Act. Marker) [GL7]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: FC; Size: 25 ug
Catalog No. 50-711-898