Learn More
Biolegend Pacific Blue™ anti-mouse Ly108
Supplier:  Biolegend 134608
Encompass_Preferred
Pacific Blue™ anti-mouse Ly108 [330-AJ]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-164-761