Learn More
Charles River Labs Cell Solutions PB CD4 CELLS- POS SEL CRYO 10M
Supplier:  Charles River Labs Cell Solutions PB04C1
Encompass
For Research Use Only. Human Normal Peripheral Blood CD4+ Helper T Cells - Cryopreserved, 10 million cells/vial. Isolated using POSITIVE immunomagnetic cell separation procedures.
Catalog No. 50-970-346
Only null left
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov