Learn More
Charles River Labs Cell Solutions Peripheral Blood CD14+ Classical Monocytes (Negative Selection), 10 Million cells, Cryopreserved
Supplier:  Charles River Labs Cell Solutions PB14NC1
Encompass
CD14+ monocytes are responsible for phagocytosis of foreign substances in the body and are capable of killing infected host cells via antibody-mediated cellular cytotoxicity. Under the right culture conditions, monocytes can differentiate into dendritic cells, Langerhans cells, or macrophages.
Catalog No. 50-225-6300
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov