Learn More
Medchemexpress LLC PB1 | 188714-28-5 | 99.5% | 296.11 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138648100MG
Encompass_Preferred
PB1 is a potent intracellular disulfide reducing agent designed with good cell permeability and the ability to form a high intracellular concentration gradient, ensuring stability. It is a borane-protected TCEP (tris(2-carboxyethyl)phosphine) analogue.
  • Potent intracellular disulfide reducing agent
  • Good cell permeability
  • Forms high intracellular concentration gradient
  • High stability
  • Borane-protected TCEP analogue
  • Increases retinal ganglion cell survival after axotomy in vitro
  • Useful for neuroprotective research
Catalog No. 50-003-50436