Learn More
Cayman Chemical PB22 3carboxyIndl metaol 10mg
SDP
Supplier:  Cayman Chemical 1438510 MG
Encompass_Preferred
An expected metabolite of PB-22 that would be detectable both in serum and urine; intended for forensic and research applications
Catalog No. 50-297-0131