Learn More
Medchemexpress LLC HY-101451 10mg Medchemexpress, PBIT CAS:2514-30-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10145110MG
Encompass_Preferred
Medchemexpress, HY-101451 10mg PBIT CAS:2514-30-9 PBIT is a specific inhibitor of the Jumonji AT-rich Interactive Domain 1 ( JARID1 ) enzymes. PBIT inhibits JARID1B ( KDM5B or PLU1 ) histone demethylase with an IC 50 of about 3 μM . PBIT also inhibits JARID1A and JARID1C with IC 50 s of 6 and 4.9 μM, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2135