Learn More
Charles River Labs Cell Solutions PBMC CRYO 50M
Supplier:  Charles River Labs Cell Solutions PB009C50
Encompass
For Research Use Only. Human Normal Peripheral Blood Mononuclear Cells (PBMC) - Cryopreserved, 50 million cells/vial. Mononuclear cells (PBMCs) are purified using density gradient centrifugation techniques.
Catalog No. 50-970-331
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov