Learn More
ABclonal Technology PBR/TSPO Rabbit pAb
SDP
Supplier:  ABclonal Technology A15649500UL
Encompass_Preferred
Present mainly in the mitochondrial compartment of peripheral tissues, the protein encoded by this gene interacts with some benzodiazepines and has different affinities than its endogenous counterpart. The protein is a key factor in the flow of cholesterol into mitochondria to permit the initiation of steroid hormone synthesis. Alternatively spliced transcript variants have been reported, one of the variants lacks an internal exon and is considered non-coding, and the other variants encode the same protein.
Catalog No. 50-001-05677