Learn More
New England Biolabs, Inc. pBR322 DNA-BstNI Digest – 0.25 ml
SDP
Supplier:  New England Biolabs, Inc. N3031L
Encompass
The BstNI Digest of pBR322 DNA yields 6 fragments suitable for use as molecular weight standards for agarose gel electrophoresis. Comes supplied with 1 vial of Gel Loading Dye, Purple (6X), no SDS.
Catalog No. 50-811-764