Learn More
Medchemexpress LLC PBRM1-BD2-IN-2 | 2819989-57-4 | C14H9Cl2FN2O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1515291MG
Encompass_Preferred
PBRM1-BD2-IN-2 is a selective and cell-active polybromo-1 (PBRM1) bromodomain inhibitor. It demonstrates binding affinity and inhibitory activity for PBRM1-BD2, making it suitable for cancer research. This compound selectively inhibits the growth of PBRM1-dependent prostate cancer cell lines.
  • Selective and cell-active polybromo-1 (PBRM1) bromodomain inhibitor
  • Has binding affinity for PBRM1-BD2 with Kd of 9.3 μM
  • Shows inhibitory activity for PBRM1-BD2 with IC50 of 1.0 μM
  • Useful for cancer research
  • Selectively inhibits the growth of PBRM1-dependent prostate cancer cell lines
Catalog No. 50-003-72359