Learn More
TARGETMOL CHEMICALS INC PBRM1-BD2-IN-3 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T601585MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PBRM1-BD2-IN-3 (compound 12) is a potent PBRM1-BD2 inhibitor with an IC50 value of 1.1 uM. PBRM1-BD2 Inhibitor can be used to research anticancer. purity: 99%
Catalog No. 50-257-9661