Learn More
Quality Biological Inc PBS (10X) pH 7.2 20L
SDP
Supplier:  Quality Biological Inc 119248161
Encompass_Preferred
PBS (phosphate buffered saline) is a balanced salt solution used for a variety biological and cell culture applications such as washing cells before dissociation transporting cells or tissue diluting cells for counting and preparing reagents.Endotoxin and Performance tested Manufactured in an ISO 90012015 certified facility.
Catalog No. 50-220-9761