Learn More
Kd Medical Inc PBS 20X PLUS TWEEN 20 4L
SDP
Supplier:  Kd Medical Inc RGH0200
Encompass_Preferred
Phosphate Buffered Saline and Tween 20 in a 20X concentrate for use as a wash buffer in immuno-labeling techniques. One 4 liter cubitainer.
Catalog No. 50-101-8908