Learn More
Boca Scientific Inc Antibody Stabilizer PBS Base, 50ml
Supplier:  Boca Scientific Inc 131 050
Encompass
Antibody Stabilizer conserves the structure of proteins and antibodies to prevent loss of functionality due to storage. With refrigerated storage, Antibody Stabilizer prevents the need for freezing and thawing. Both TRIS-based and PBS-based Antibody Stabilizers are available, allowing the user to select the optimal buffer base for the assay.
Catalog No. NC0436689