Learn More
Crystalgen Inc PBS PH7.2 10X 12 X 500ML
SDP
Supplier:  Crystalgen Inc 22114112CS12
Encompass
PBS (Phosphate Buffered Saline) pH7.2 10X, with Ca, Mg-500mlUltra pure Phosphate buffered isotonic saline. With calcium and magnesium. Sterile filtered. 1X PBS solutions contains 137mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 2mM KH2PO4.pH: 7.2Storage: 18-26oC
Catalog No. NC2302963