Learn More
Research Products International Corp PBS/Tween 20 10X Solution, 500 Milliliters
SDP
Supplier:  Research Products International Corp P21200500.0
Encompass_Preferred
PBS/Tween 20 10X Solution is a widely used buffer solution in the life science marketplace. It is a combination of phosphate-buffered saline (PBS) and Tween 20, a nonionic detergent.
Catalog No. 50-136-8500