Learn More
Alkali Scientific PBS/Tween 20 10X Solution, 500 Milliliters
SDP
Supplier:  Alkali Scientific BST001CP
Encompass_Preferred
PBS/Tween 20 10X Solution is a concentrated buffer solution that combines the benefits of phosphate-buffered saline with Tween 20, a non-ionic detergent, to enhance the solubilization of biological molecules. This 500mL solution is commonly used for washing and blocking steps in immunological assays, particularly in enzyme-linked immunosorbent assays (ELISA) and western blotting. The addition of Tween 20 helps to reduce non-specific binding and improve the sensitivity of the assay. The 10X concentration allows for easy dilution to working strength, making this product versatile for various laboratory applications. It is an essential reagent for researchers working with proteins, antibodies, and cell cultures.
Catalog No. 50-006-60698