Learn More
Medchemexpress LLC PC alkyne-PEG4-NHS ester | 1802907-98-7 | 95.0% | 653.63 g/mol | C29H39N3O14 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY14013925MG
Encompass_Preferred
PC alkyne-PEG4-NHS ester is a cleavable PEG4 N-hydroxysuccinimidyl (NHS) ester bearing an alkyne handle for click chemistry, used for bioconjugation and as an antibody-drug conjugate (ADC) linker. It reacts with amines via the NHS ester and undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAC) through the terminal alkyne.
  • Cleavable four-unit PEG linker suitable for bioconjugation.
  • NHS ester functional group for efficient amine coupling.
  • Alkyne handle enables CuAAC (click chemistry) with azides.
  • Typical purity around 95.0% for research use.
  • Molecular weight approximately 653.63 g/mol; viscous light-yellow appearance.
  • Commonly used in antibody-drug conjugate synthesis and other bioconjugation workflows.
Catalog No. 50-002-03757