Learn More
Medchemexpress LLC PC biotin-PEG3-azide | 1937270-46-6 | 97.8% | 825.93 | C35H55N9O12S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1401325MG
Encompass_Preferred
PC Biotin-PEG3-azide is a cleavable three-unit polyethylene glycol linker containing a biotin tag and an azide functional group. It is used for antibody-drug conjugate synthesis and bioconjugation via copper-catalyzed or strain-promoted azide-alkyne cycloaddition.
  • Cleavable three-unit PEG linker for ADC and conjugation chemistry.
  • Azide functional group compatible with CuAAC and SPAAC reactions.
  • Biotin tag enables detection and affinity capture.
  • High chemical purity suitable for research applications.
  • Available in small research quantities (5 mg, 10 mg, larger by quote).
Catalog No. 50-002-03810