Learn More
Medchemexpress LLC PC-Biotin-PEG4-NHS carbonate | 2055198-03-1 | 810.87 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14013550MG
Encompass_Preferred
PC-Biotin-PEG4-NHS carbonate is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker; one ligand is for an E3 ubiquitin ligase and the other is for the target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51379