Learn More
Medchemexpress LLC PC-Biotin-PEG4-NHS carbonate | 2055198-03-1 | MFCD29079395 | > 96% | 810.87 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY140135
Encompass_Preferred
PC-Biotin-PEG4-NHS carbonate is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a unique amine-reactive, photocleavable biotin reagent, making it useful for introducing a biotin moiety to amine-containing molecules. This compound also serves as a linker for bio-conjugation, featuring a terminal NHS carbonate and a photocleavable biotin group, all linked through a linear PEG chain.
  • PEG-based PROTAC linker
  • Amine reactive
  • Photocleavable biotin reagent
  • Introduces biotin moiety to amine-containing molecules
  • Linker for bio-conjugation
  • Contains a terminal NHS carbonate
  • Linked through a linear PEG chain
Catalog No. 50-003-49414