Learn More
Grainger PC PRODUCTS, Epoxy Adhesive, PC Clear
Supplier:  Grainger 25F529
Encompass_Preferred
PC Products PC-Clear Epoxy Adhesive is a two-part epoxy that creates strong, clear bonds on metals, glass, ceramics, wood, and most plastics. It has a working time of 25-30 minutes and reaches handling strength in 2-3 hours at room temperature. This adhesive can fill gaps up to 1/4 inch and has excellent resistance to chemicals and weathering. It comes in a dual syringe with static mixer tip for easy dispensing and mixing. This versatile epoxy is ideal for both small repairs and large projects.
Catalog No. NC2602169