Learn More
Medchemexpress LLC PC Mal-NHS carbonate ester | 1408057-91-9 | 95.0% | 562.48 | C24H26N4O12 | 10 MG

Supplier: Medchemexpress LLC HY14014010MG
PC Mal-NHS carbonate ester is a photocleavable cleavable linker reagent designed for bioconjugation in the synthesis of antibody-drug conjugates (ADCs). It provides maleimide and NHS carbonate reactive groups for thiol and amine coupling and is supplied as a stable solid for research use.
- Photocleavable linker for controlled release in ADCs.
- Contains maleimide and NHS carbonate reactive groups.
- White to off-white solid powder suitable for storage at low temperature.
- Molecular weight 562.48 and chemical formula C24H26N4O12.
- High purity (95.0%) for research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.