Learn More
Echelon Biosciences Research Labs PC/PS LIPID MICROPARTICL 100UL
SDP
Supplier:  Echelon Biosciences Research Labs PB1PCPS100UL
Encompass
PC/PS Lipid Microparticles contain attached phosphatidylcholine and phosphatidylserine with the phospholipid headgroups exposed and available for biological interactions. Phosphatidylserine (PS) is an anionic, intracellular phospholipid component of the cell membrane and is involved in cell signaling including apoptosis. Phosphatidylcholine (PC) is primarily an extracellular phospholipid component of cell membranes and is a key structural lipid. This item is non returnable
Catalog No. NC1972397