Learn More
Werther International PC106 SILENT COMPRESSOR 115V
SDP
Supplier:  Werther International M8037US
Encompass_Preferred
Quiet oil free compressor, PC106 115V/60Hz, Output 4.4 CFM / 110 l.min, Max Pressure 120 PSI, Noise Level 57 dB/A, 1.57 Gal air tank, 2 year warranty, Dimensions 24 in.L x 14 in.W x 23 in.H
Catalog No. 50-103-7818