Learn More
Enzo Life Sciences PC2 polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2690100
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 87-116 of N-terminal human PC2 conjugated to KLH. Species reactivity: Human, Mouse. Applications: Flow Cytometry, ICC, IF, IHC, WB. Formulation: Liquid. In PBS containing 0.09% sodium azide.
Catalog No. 50-201-2814