Learn More
Medchemexpress LLC PC786 - RSV L inhibitor | Purity: 99.69% | Mol Wt: 715.83 | 1902114-15-1 | 5mg
SDP
Supplier:  Medchemexpress LLC HY1020385MG
Encompass_Preferred
PC786 - RSV L inhibitor | Purity: 99.69% | Mol Wt: 715.83 | 1902114-15-1 | 5mg
Catalog No. NC2104668