Learn More
Medchemexpress LLC HY-117766 5mg Medchemexpress, PC945 CAS:1931946-73-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1177665MG
Encompass_Preferred
Medchemexpress, HY-117766 5mg PC945 CAS:1931946-73-4 PC945, a potent, long-acting antifungal triazole, possesses activity against a broad range of both azole-susceptible and azole-resistant strains of Aspergillus fumigatus. PC945 is also a potent, tightly binding inhibitor of A. fumigatus sterol 14α-demethylase activity, CYP51A and CYP51B, with IC50s of 0.23 μM and 0.22 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0909