Learn More
Electron Microscopy Sciences PCF Lasner-Lamann Tissue Forceps with 5"
SDP
Supplier:  Electron Microscopy Sciences 77893LM2.5
Encompass_Preferred
Pressure controlled Lasner-DaMan Tissue Forceps virtually eliminates the penetration or crushing of fragile tissue while providing uncompromised slip resistance. The ergonomic Featherlite spring handle relieves excess pressure and mechanical stress for ideal compression in either hand. The patented twisted I-beam design with three point fixation provides unparralled stability and micro cross-cut carbide jaws provide an extraordinary grip on tissue and suture needles. Adjustable auto-lock feature permits auto-retention of tissue.
Catalog No. 50-306-39