Learn More
Med Vet International Dynarex Sterilization Pouches, 2.25" X 2.75", 5000/Case
Supplier:  Med Vet International 4460
Encompass_Preferred
Dynarex Sterilization Pouches are high-quality, self-sealing pouches designed to facilitate the sterilization of medical instruments. These Pouches are crafted from tear-resistant medical paper for added durability. The Sterilization Pouches feature blue-tinted transparent film for easy detection of punctures or tears. The pouches also feature color-changing indicators to help ensure proper sterilization levels.  Dynarex Sterilization Pouches are premium-quality pouches crafted from tear-resistant medical paper for use during the sterilization of medical instruments. These self-sealing Pouches feature a blue-tinted transparent film to easily detect punctures and tears. 
Catalog No. 50-282-8949