Learn More
TARGETMOL CHEMICALS INC PCI-34051 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T632525MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PCI-34051 is an effective and selective HDAC8 inhibitor (IC50 10 nM). purity: 99%
Catalog No. 50-257-9431