Learn More
ABclonal Technology PCK1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A22172100UL
Encompass_Preferred
This gene is a main control point for the regulation of gluconeogenesis The cytosolic enzyme encoded by this gene along with GTP catalyzes the formation of phosphoenolpyruvate from oxaloacetate with the release of carbon dioxide and GDP The expression of this gene can be regulated by insulin glucocorticoids glucagon cAMP and diet Defects in this gene are a cause of cytosolic phosphoenolpyruvate carboxykinase deficiency A mitochondrial isozyme of the encoded protein also has been characterized
Catalog No. 50-000-07163